Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAAAGACACCGAGATTCAAGCC[A/T]TCTAGTAGGTTGAGGACGCTGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604908 MIM: 605801 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP4R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP4R1 - protein phosphatase 4 regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042388.2 | 2968 | UTR 3 | NP_001035847.1 | |||
NM_005134.3 | 2968 | UTR 3 | NP_005125.1 | |||
XM_011525775.2 | 2968 | UTR 3 | XP_011524077.1 | |||
XM_011525776.1 | 2968 | UTR 3 | XP_011524078.1 | |||
XM_011525779.2 | 2968 | UTR 3 | XP_011524081.1 | |||
XM_017026105.1 | 2968 | UTR 3 | XP_016881594.1 | |||
XM_017026106.1 | 2968 | UTR 3 | XP_016881595.1 | |||
XM_017026107.1 | 2968 | UTR 3 | XP_016881596.1 |
RALBP1 - ralA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |