Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCCTAGTGGCATTTGCCACTG[C/G]AAGTGGAAGCATTTAACAAGGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612135 MIM: 606730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABYR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABYR - calcium binding tyrosine phosphorylation regulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308231.1 | 4165 | Intron | NP_001295160.1 | |||
NM_012189.3 | 4165 | Intron | NP_036321.2 | |||
NM_138643.2 | 4165 | Intron | NP_619584.1 | |||
NM_138644.2 | 4165 | Intron | NP_619585.1 | |||
NM_153768.2 | 4165 | Intron | NP_722452.1 | |||
NM_153769.2 | 4165 | Intron | NP_722453.1 | |||
NM_153770.2 | 4165 | Intron | NP_722454.1 | |||
XM_005258247.2 | 4165 | Intron | XP_005258304.1 | |||
XM_017025706.1 | 4165 | Intron | XP_016881195.1 |
OSBPL1A - oxysterol binding protein like 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242508.1 | 4165 | UTR 3 | NP_001229437.1 | |||
NM_018030.4 | 4165 | UTR 3 | NP_060500.3 | |||
NM_080597.3 | 4165 | UTR 3 | NP_542164.2 | |||
XM_006722380.3 | 4165 | UTR 3 | XP_006722443.1 | |||
XM_006722382.3 | 4165 | UTR 3 | XP_006722445.1 | |||
XM_017025530.1 | 4165 | UTR 3 | XP_016881019.1 | |||
XM_017025531.1 | 4165 | UTR 3 | XP_016881020.1 | |||
XM_017025532.1 | 4165 | Intron | XP_016881021.1 | |||
XM_017025533.1 | 4165 | UTR 3 | XP_016881022.1 | |||
XM_017025534.1 | 4165 | UTR 3 | XP_016881023.1 |