Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTCACCCAAGGTCAAGGCCAG[A/G]AAGTGGCATCTCTGGATGCCCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607320 MIM: 180901 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASGRP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASGRP4 - RAS guanyl releasing protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RYR1 - ryanodine receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000540.2 | Intron | NP_000531.2 | ||||
NM_001042723.1 | Intron | NP_001036188.1 | ||||
XM_006723317.2 | Intron | XP_006723380.1 | ||||
XM_006723319.2 | Intron | XP_006723382.1 | ||||
XM_011527205.2 | Intron | XP_011525507.1 |