Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTTTTAAATTTTGAAACATTGC[A/G]TAATATAAAAATGTTTTTATTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602102 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC723805 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC723805 - interleukin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUPT5H - SPT5 homolog, DSIF elongation factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111020.2 | Intron | NP_001104490.1 | ||||
NM_001130824.1 | Intron | NP_001124296.1 | ||||
NM_001130825.1 | Intron | NP_001124297.1 | ||||
NM_001319990.1 | Intron | NP_001306919.1 | ||||
NM_001319991.1 | Intron | NP_001306920.1 | ||||
NM_003169.3 | Intron | NP_003160.2 | ||||
XM_017027174.1 | Intron | XP_016882663.1 |