Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGAGTGTCCCAGTAAAGGTTAAC[A/G]CTGATCAGGTAAACCGGGGGTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OPA3 - optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017989.2 | 964 | Intron | NP_001017989.2 | |||
NM_025136.3 | 964 | UTR 3 | NP_079412.1 | |||
XM_006723403.3 | 964 | UTR 3 | XP_006723466.1 |