Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCGCGATAGCAGGGCTGGGTGTT[A/G]TCACCCCAGTAAACCAGTGTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601991 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOVA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOVA2 - NOVA alternative splicing regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002516.3 | 5961 | UTR 3 | NP_002507.1 | |||
XM_006723230.3 | 5961 | Intron | XP_006723293.1 | |||
XM_017026838.1 | 5961 | Intron | XP_016882327.1 | |||
XM_017026839.1 | 5961 | Intron | XP_016882328.1 | |||
XM_017026840.1 | 5961 | Intron | XP_016882329.1 |