Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTCCTCCAGTGCTCTGGCTAT[C/T]GCTGAAAACACCTTCTAGTTCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603326 MIM: 604105 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM217B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM217B - family with sequence similarity 217 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190826.1 | 2296 | UTR 5 | NP_001177755.1 | |||
NM_001190827.1 | 2296 | Intron | NP_001177756.1 | |||
NM_022106.2 | 2296 | Intron | NP_071389.1 | |||
XM_011528985.2 | 2296 | Intron | XP_011527287.1 | |||
XM_011528986.2 | 2296 | UTR 5 | XP_011527288.1 |
PPP1R3D - protein phosphatase 1 regulatory subunit 3D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006242.3 | 2296 | UTR 3 | NP_006233.1 |
SYCP2 - synaptonemal complex protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |