Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGAATAACTACTCAGAAGGTGC[C/T]GTGGACTGAAAGTGCGTATGGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603152 MIM: 600609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5J PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5J - ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GABPA - GA binding protein transcription factor alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197297.1 | 639 | Intron | NP_001184226.1 | |||
NM_002040.3 | 639 | Intron | NP_002031.2 | |||
XM_005260938.4 | 639 | Intron | XP_005260995.1 | |||
XM_011529520.2 | 639 | Intron | XP_011527822.1 | |||
XM_011529521.2 | 639 | UTR 5 | XP_011527823.1 | |||
XM_017028313.1 | 639 | Intron | XP_016883802.1 |