Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCAAGCCCGGCTGCCCTTCGGT[A/G]CCACCAGCACCACTGCTCCGTCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETS2 - ETS proto-oncogene 2, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256295.1 | 95 | Missense Mutation | ACC,GCC | T,A 19 | NP_001243224.1 | |
NM_005239.5 | 95 | Intron | NP_005230.1 | |||
XM_005260935.1 | 95 | Intron | XP_005260992.1 | |||
XM_017028290.1 | 95 | Intron | XP_016883779.1 |