Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCTAAAAGTAGTAAAACTTCAG[A/G]ACCTTCTTCTTCCCCAGTTCTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300889 MIM: 300897 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H12B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3H12B - zinc finger CCCH-type containing 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010888.3 | Intron | NP_001010888.3 | ||||
XM_011530939.2 | Intron | XP_011529241.1 | ||||
XM_017029478.1 | Intron | XP_016884967.1 | ||||
XM_017029479.1 | Intron | XP_016884968.1 | ||||
XM_017029480.1 | Intron | XP_016884969.1 | ||||
XM_017029481.1 | Intron | XP_016884970.1 | ||||
XM_017029482.1 | Intron | XP_016884971.1 | ||||
XM_017029483.1 | Intron | XP_016884972.1 | ||||
XM_017029484.1 | Intron | XP_016884973.1 |
ZC4H2 - zinc finger C4H2-type containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |