Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATATCAATGAAGACAGTAACTT[A/C]TTAGGAAGACAGGATTATTAAAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300196 | ||||||||||||||||||||
Literature Links: |
TBL1X PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBL1X - transducin (beta)-like 1X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139466.1 | Intron | NP_001132938.1 | ||||
NM_001139467.1 | Intron | NP_001132939.1 | ||||
NM_001139468.1 | Intron | NP_001132940.1 | ||||
NM_005647.3 | Intron | NP_005638.1 | ||||
XM_011545571.2 | Intron | XP_011543873.1 |