Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTCTTAGATCATTTATCAATCA[A/G]AGGAAACAGACTGGAAGTCAAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELOVL6 - ELOVL fatty acid elongase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130721.1 | Intron | NP_001124193.1 | ||||
NM_024090.2 | Intron | NP_076995.1 | ||||
XM_011532233.2 | Intron | XP_011530535.1 | ||||
XM_011532234.2 | Intron | XP_011530536.1 | ||||
XM_011532235.2 | Intron | XP_011530537.1 |