Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTGTGCTGCTGCTTCTAAAATA[G/A]GAACAAGGACAAAAATCATTTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603148 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF3 - activating transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030287.3 | Intron | NP_001025458.1 | ||||
NM_001040619.2 | Intron | NP_001035709.1 | ||||
NM_001206484.2 | Intron | NP_001193413.2 | ||||
NM_001206486.2 | Intron | NP_001193415.1 | ||||
NM_001206488.2 | Intron | NP_001193417.2 | ||||
NM_001674.3 | Intron | NP_001665.1 | ||||
XM_005273146.1 | Intron | XP_005273203.1 | ||||
XM_011509579.1 | Intron | XP_011507881.1 |