Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCTCTCTTCTTTGCCGTGACTG[C/T]CCCAGCCAAGCCACTCTCCCCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSD4 - pleckstrin and Sec7 domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012455.2 | Intron | NP_036587.2 | ||||
XM_005263634.2 | Intron | XP_005263691.1 | ||||
XM_006712392.2 | Intron | XP_006712455.1 | ||||
XM_006712393.2 | Intron | XP_006712456.1 | ||||
XM_006712394.2 | Intron | XP_006712457.1 |