Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAATGCTATTCTAATTAAGAGAG[C/T]ATATGATGAAAAATATGTGAGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSIG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INSIG2 - insulin induced gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321329.1 | Intron | NP_001308258.1 | ||||
NM_001321330.1 | Intron | NP_001308259.1 | ||||
NM_001321331.1 | Intron | NP_001308260.1 | ||||
NM_001321332.1 | Intron | NP_001308261.1 | ||||
NM_001321333.1 | Intron | NP_001308262.1 | ||||
NM_016133.3 | Intron | NP_057217.2 |