Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAAGACAAAGGCTCCACTCCC[A/G]TGGTGCTTACATTCTGGCAGGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZAP70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZAP70 - zeta chain of T cell receptor associated protein kinase 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079.3 | Intron | NP_001070.2 | ||||
NM_207519.1 | Intron | NP_997402.1 | ||||
XM_017004867.1 | Intron | XP_016860356.1 | ||||
XM_017004868.1 | Intron | XP_016860357.1 | ||||
XM_017004869.1 | Intron | XP_016860358.1 | ||||
XM_017004870.1 | Intron | XP_016860359.1 |