Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAAATATTCTGGCCTCATCTATC[C/T]CTGGTTCACTTGTCAAGGCTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603706 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
M1AP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
M1AP - meiosis 1 associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA4F - ssemaphorin 4F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271661.1 | Intron | NP_001258590.1 | ||||
NM_001271662.1 | Intron | NP_001258591.1 | ||||
NM_004263.4 | Intron | NP_004254.2 | ||||
XM_011532475.2 | Intron | XP_011530777.1 | ||||
XM_011532476.2 | Intron | XP_011530778.1 | ||||
XM_011532477.2 | Intron | XP_011530779.1 | ||||
XM_017003172.1 | Intron | XP_016858661.1 | ||||
XM_017003173.1 | Intron | XP_016858662.1 | ||||
XM_017003174.1 | Intron | XP_016858663.1 |