Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAGCATGGCCCTTGTGCCAAGC[C/T]CATAATGATAGTCCTATGGGATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM149A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM149A - family with sequence similarity 149 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006655.2 | Intron | NP_001006656.1 | ||||
NM_015398.2 | Intron | NP_056213.1 | ||||
XM_005262911.1 | Intron | XP_005262968.1 | ||||
XM_005262912.1 | Intron | XP_005262969.1 | ||||
XM_005262913.1 | Intron | XP_005262970.1 | ||||
XM_011531834.1 | Intron | XP_011530136.1 | ||||
XM_011531835.2 | Intron | XP_011530137.1 | ||||
XM_011531836.1 | Intron | XP_011530138.1 | ||||
XM_017007996.1 | Intron | XP_016863485.1 | ||||
XM_017007997.1 | Intron | XP_016863486.1 | ||||
XM_017007998.1 | Intron | XP_016863487.1 | ||||
XM_017007999.1 | Intron | XP_016863488.1 | ||||
XM_017008000.1 | Intron | XP_016863489.1 | ||||
XM_017008001.1 | Intron | XP_016863490.1 | ||||
XM_017008002.1 | Intron | XP_016863491.1 | ||||
XM_017008003.1 | Intron | XP_016863492.1 | ||||
XM_017008004.1 | Intron | XP_016863493.1 | ||||
XM_017008005.1 | Intron | XP_016863494.1 | ||||
XM_017008006.1 | Intron | XP_016863495.1 | ||||
XM_017008007.1 | Intron | XP_016863496.1 | ||||
XM_017008008.1 | Intron | XP_016863497.1 |