Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTATGGCTTTCCTCATGCTGCC[C/T]GGAAGCTACTTCCCCATAAAGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601459 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNOC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNOC - prepronociceptin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284244.1 | Intron | NP_001271173.1 | ||||
NM_006228.4 | Intron | NP_006219.1 | ||||
XM_005273532.2 | Intron | XP_005273589.1 | ||||
XM_011544559.2 | Intron | XP_011542861.1 | ||||
XM_017013578.1 | Intron | XP_016869067.1 |