Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGCCCCTCAGGTCATAGTGA[A/G]ACAAGTCTGCTGCTGCGTGGCGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBRSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBRSL1 - fibrosin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142641.1 | Intron | NP_001136113.1 | ||||
XM_005266171.3 | Intron | XP_005266228.1 | ||||
XM_005266173.3 | Intron | XP_005266230.1 | ||||
XM_005266175.3 | Intron | XP_005266232.1 | ||||
XM_005266176.3 | Intron | XP_005266233.1 | ||||
XM_005266177.3 | Intron | XP_005266234.1 | ||||
XM_005266181.3 | Intron | XP_005266238.1 | ||||
XM_011534803.2 | Intron | XP_011533105.1 | ||||
XM_011534804.2 | Intron | XP_011533106.1 | ||||
XM_011534805.2 | Intron | XP_011533107.1 | ||||
XM_011534806.2 | Intron | XP_011533108.1 | ||||
XM_011534807.2 | Intron | XP_011533109.1 | ||||
XM_011534808.2 | Intron | XP_011533110.1 | ||||
XM_011534809.2 | Intron | XP_011533111.1 | ||||
XM_011534810.2 | Intron | XP_011533112.1 | ||||
XM_011534812.2 | Intron | XP_011533114.1 | ||||
XM_011534816.2 | Intron | XP_011533118.1 |