Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCATAGTGCTTCACCATCCCA[A/G]TGGGAATGGTAAAGATGGAAGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616909 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC68 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC68 - coiled-coil domain containing 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143829.1 | Intron | NP_001137301.1 | ||||
NM_025214.2 | Intron | NP_079490.1 | ||||
XM_006722552.3 | Intron | XP_006722615.1 | ||||
XM_011526198.2 | Intron | XP_011524500.1 | ||||
XM_011526199.2 | Intron | XP_011524501.1 | ||||
XM_011526200.2 | Intron | XP_011524502.1 | ||||
XM_011526201.2 | Intron | XP_011524503.1 | ||||
XM_011526202.1 | Intron | XP_011524504.1 | ||||
XM_011526203.2 | Intron | XP_011524505.1 | ||||
XM_011526204.2 | Intron | XP_011524506.1 | ||||
XM_017026011.1 | Intron | XP_016881500.1 |