Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATATTTTGCTTTAAATGTGCAGG[G/T]TAAAGTGTTATAATTCCTTCTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605521 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBPL1 - TATA-box binding protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253676.1 | Intron | NP_001240605.1 | ||||
NM_004865.3 | Intron | NP_004856.1 | ||||
XM_017011513.1 | Intron | XP_016867002.1 |