Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCCTGGTACAGTCACCGACTG[C/T]GTGTAGCCCACTCTGAAAACGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT16 - polypeptide N-acetylgalactosaminyltransferase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168368.1 | Intron | NP_001161840.1 | ||||
NM_020692.2 | Intron | NP_065743.2 | ||||
XM_011537005.1 | Intron | XP_011535307.1 | ||||
XM_011537006.2 | Intron | XP_011535308.1 | ||||
XM_011537007.2 | Intron | XP_011535309.1 | ||||
XM_017021498.1 | Intron | XP_016876987.1 |
LOC105370550 - uncharacterized LOC105370550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |