Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGATAATCACATTTAAAAATGTA[C/T]GTTTTCCTCTTTCAAAAAAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607047 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATXN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATXN3 - ataxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127696.1 | 5905 | UTR 3 | NP_001121168.1 | |||
NM_001127697.2 | 5905 | UTR 3 | NP_001121169.2 | |||
NM_001164774.1 | 5905 | UTR 3 | NP_001158246.1 | |||
NM_001164776.1 | 5905 | UTR 3 | NP_001158248.1 | |||
NM_001164777.1 | 5905 | UTR 3 | NP_001158249.1 | |||
NM_001164778.1 | 5905 | UTR 3 | NP_001158250.1 | |||
NM_001164779.1 | 5905 | UTR 3 | NP_001158251.1 | |||
NM_001164780.1 | 5905 | UTR 3 | NP_001158252.1 | |||
NM_001164781.1 | 5905 | UTR 3 | NP_001158253.1 | |||
NM_001164782.1 | 5905 | UTR 3 | NP_001158254.1 | |||
NM_004993.5 | 5905 | UTR 3 | NP_004984.2 | |||
NM_030660.4 | 5905 | UTR 3 | NP_109376.1 |