Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTAGTAGTCCTCCCTTATCCA[G/T]GGTTTTGCTTTCTGCAGTCTCAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616790 MIM: 605271 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP4R4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP4R4 - protein phosphatase 4 regulatory subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINA10 - serpin family A member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100607.2 | 2402 | UTR 3 | NP_001094077.1 | |||
NM_016186.2 | 2402 | UTR 3 | NP_057270.1 | |||
XM_005267733.4 | 2402 | UTR 3 | XP_005267790.1 | |||
XM_017021353.1 | 2402 | UTR 3 | XP_016876842.1 |