Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCTGTATAAAATATTCAATTCC[A/G]TTGAAGTGAGCCTACAAAAAATATG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612688 | |||||||||||||||||||||||
Literature Links: |
RNF168 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RNF168 - ring finger protein 168 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMCO1 - single-pass membrane protein with coiled-coil domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077657.2 | 369 | Missense Mutation | ACG,ATG | T,M 71 | NP_001071125.1 | |
NM_001320473.1 | 369 | Missense Mutation | ACG,ATG | T,M 63 | NP_001307402.1 |