Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAATCCATCCTCCATTAAGGCT[A/G]TCAGCACTTTGGGGTGCCTGCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606076 MIM: 613170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3R3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3R3 - phosphoinositide-3-kinase regulatory subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114172.1 | Intron | NP_001107644.1 | ||||
NM_001303427.1 | Intron | NP_001290356.1 | ||||
NM_001303428.1 | Intron | NP_001290357.1 | ||||
NM_001303429.1 | Intron | NP_001290358.1 | ||||
NM_003629.3 | Intron | NP_003620.3 | ||||
XM_017002614.1 | Intron | XP_016858103.1 |
TSPAN1 - tetraspanin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005727.3 | Intron | NP_005718.2 | ||||
XM_011540460.2 | Intron | XP_011538762.1 |