Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTACATAGACACCATGCTCTGT[C/T]CCCACCCCAATCTGCCAGCCCTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611001 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEAF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEAF6 - MYST/Esa1 associated factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270875.1 | Intron | NP_001257804.1 | ||||
NM_001270876.1 | Intron | NP_001257805.1 | ||||
NM_022756.5 | Intron | NP_073593.2 |
MIR5581 - microRNA 5581 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |