Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCATAGAGTAAACTCTACTTCAC[A/G]GAGCTCCCAGTTGAGTGTGATAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600282 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIK4 - glutamate ionotropic receptor kainate type subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282470.2 | Intron | NP_001269399.1 | ||||
NM_001282473.2 | Intron | NP_001269402.1 | ||||
NM_014619.4 | Intron | NP_055434.2 | ||||
XM_011542784.1 | Intron | XP_011541086.1 | ||||
XM_011542786.2 | Intron | XP_011541088.1 | ||||
XM_011542787.2 | Intron | XP_011541089.1 | ||||
XM_017017621.1 | Intron | XP_016873110.1 | ||||
XM_017017622.1 | Intron | XP_016873111.1 |