Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTCTCCATGATATTTGCTACT[C/T]GAGTTGAGGGTAAAAAGCTAATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609317 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM36 - tripartite motif containing 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017397.1 | 3901 | Intron | NP_001017397.1 | |||
NM_001017398.1 | 3901 | Intron | NP_001017398.1 | |||
NM_001300752.1 | 3901 | UTR 3 | NP_001287681.1 | |||
NM_001300759.1 | 3901 | UTR 3 | NP_001287688.1 | |||
NM_018700.3 | 3901 | UTR 3 | NP_061170.2 | |||
XM_017009621.1 | 3901 | Intron | XP_016865110.1 | |||
XM_017009622.1 | 3901 | Intron | XP_016865111.1 | |||
XM_017009623.1 | 3901 | Intron | XP_016865112.1 |