Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTGCCACTCACTAGCATGTGA[C/T]CCTGACAAATCATTTCACCTCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606938 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UROS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UROS - uroporphyrinogen III synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000375.2 | Intron | NP_000366.1 | ||||
NM_001324036.1 | Intron | NP_001310965.1 | ||||
NM_001324037.1 | Intron | NP_001310966.1 | ||||
NM_001324038.1 | Intron | NP_001310967.1 | ||||
NM_001324039.1 | Intron | NP_001310968.1 | ||||
XM_005270140.4 | Intron | XP_005270197.1 | ||||
XM_011540127.1 | Intron | XP_011538429.1 | ||||
XM_017016610.1 | Intron | XP_016872099.1 | ||||
XM_017016611.1 | Intron | XP_016872100.1 | ||||
XM_017016612.1 | Intron | XP_016872101.1 |