Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTATAGCACTGACTCTGATGGA[C/T]AAAACGGATTTTTCTCCCTCATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UPF2 - UPF2 regulator of nonsense transcripts homolog (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015542.3 | 4555 | UTR 3 | NP_056357.1 | |||
NM_080599.2 | 4555 | UTR 3 | NP_542166.1 | |||
XM_011519449.2 | 4555 | Intron | XP_011517751.1 | |||
XM_017016034.1 | 4555 | UTR 3 | XP_016871523.1 |