Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACCTTCCTAGGTTTACATGGTA[C/T]CGGAGCCAGCAGCCTGGTGAGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF16 - Rho guanine nucleotide exchange factor 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEGF6 - multiple EGF like domains 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001409.3 | 5056 | UTR 3 | NP_001400.3 | |||
XM_006710406.3 | 5056 | UTR 3 | XP_006710469.1 | |||
XM_011540885.1 | 5056 | UTR 3 | XP_011539187.1 | |||
XM_011540886.1 | 5056 | UTR 3 | XP_011539188.1 | |||
XM_011540887.2 | 5056 | UTR 3 | XP_011539189.1 | |||
XM_011540888.2 | 5056 | Intron | XP_011539190.1 | |||
XM_017000533.1 | 5056 | UTR 3 | XP_016856022.1 |