Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCCGGCCCCCGGGGCCCTCTG[C/T]TTAGGCTGGGGACACCAGGCCTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf204 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf204 - chromosome 1 open reading frame 204 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134233.1 | 1704 | Intron | NP_001127705.1 |
VSIG8 - V-set and immunoglobulin domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013661.1 | 1704 | UTR 3 | NP_001013683.1 |