Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACTCCTATACAAAGGAAACTTT[A/G]GAGAAAGCCTCATAAAATGTTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL4-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTSL4-AS1 - ADAMTSL4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCL1 - BCL2 family apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197320.1 | 2902 | UTR 3 | NP_001184249.1 | |||
NM_021960.4 | 2902 | UTR 3 | NP_068779.1 | |||
NM_182763.2 | 2902 | UTR 3 | NP_877495.1 |