Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAAAGTCAAAAGTTAAATATTA[G/T]CCTATAAACATTTCAGGAAAAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC9C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC9C2 - solute carrier family 9 member C2 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178527.3 | Intron | NP_848622.2 | ||||
XM_011509426.1 | Intron | XP_011507728.1 | ||||
XM_017001065.1 | Intron | XP_016856554.1 | ||||
XM_017001066.1 | Intron | XP_016856555.1 | ||||
XM_017001067.1 | Intron | XP_016856556.1 | ||||
XM_017001068.1 | Intron | XP_016856557.1 | ||||
XM_017001069.1 | Intron | XP_016856558.1 | ||||
XM_017001070.1 | Intron | XP_016856559.1 | ||||
XM_017001071.1 | Intron | XP_016856560.1 | ||||
XM_017001072.1 | Intron | XP_016856561.1 | ||||
XM_017001073.1 | Intron | XP_016856562.1 | ||||
XM_017001074.1 | Intron | XP_016856563.1 | ||||
XM_017001075.1 | Intron | XP_016856564.1 | ||||
XM_017001076.1 | Intron | XP_016856565.1 |