Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTTTCTAACTTCTGGAGGTTA[C/T]CTGCATTCTAATTTGCAGCCCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608985 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF2 - ring finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007212.3 | Intron | NP_009143.1 | ||||
XM_005245413.3 | Intron | XP_005245470.1 | ||||
XM_011509851.2 | Intron | XP_011508153.1 | ||||
XM_011509852.2 | Intron | XP_011508154.1 |