Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGGCCTGCAGGAGCCAGATTG[C/T]GGCCCCCAGGGAAAACCCAGGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHIT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHIT1 - chitinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256125.1 | 1525 | UTR 3 | NP_001243054.2 | |||
NM_001270509.1 | 1525 | UTR 3 | NP_001257438.1 | |||
NM_003465.2 | 1525 | UTR 3 | NP_003456.1 |