Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAGAGGGTTTCACACAGAAACA[C/T]TCTGGGGGCCTTCGGTGACAAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHDC8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHDC8A - kelch domain containing 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271863.1 | 2365 | UTR 3 | NP_001258792.1 | |||
NM_001271864.1 | 2365 | UTR 3 | NP_001258793.1 | |||
NM_001271865.1 | 2365 | UTR 3 | NP_001258794.1 | |||
NM_018203.2 | 2365 | UTR 3 | NP_060673.1 |