Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGGAGGGGGCTGACACTCTAC[C/T]TGGGCTCAGGAAAGTGTTGCTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIRREL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIRREL - kin of IRRE like (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286349.1 | Intron | NP_001273278.1 | ||||
NM_018240.6 | Intron | NP_060710.3 | ||||
XM_005245305.4 | Intron | XP_005245362.1 |