Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACCAAATTACAACTGAAAACCAA[A/T]AATAATTTAACAGAAAATGCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHN1 - chimerin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025201.3 | 2364 | UTR 3 | NP_001020372.2 | |||
NM_001206602.1 | 2364 | UTR 3 | NP_001193531.1 | |||
NM_001822.5 | 2364 | UTR 3 | NP_001813.1 |