Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCAGTGTGCTAGGAATGGCCTC[A/G]CTGACAGTTGGAATCACGGATACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHM3 - pleckstrin homology domain containing M3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080475.2 | 4996 | UTR 3 | NP_001073944.1 | |||
XM_005246545.3 | 4996 | UTR 3 | XP_005246602.1 | |||
XM_006712521.3 | 4996 | Intron | XP_006712584.1 | |||
XM_011511160.2 | 4996 | UTR 3 | XP_011509462.1 | |||
XM_011511163.2 | 4996 | Intron | XP_011509465.1 | |||
XM_017004071.1 | 4996 | Intron | XP_016859560.1 | |||
XM_017004072.1 | 4996 | UTR 3 | XP_016859561.1 | |||
XM_017004073.1 | 4996 | UTR 3 | XP_016859562.1 |