Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAGGACATTCGTAGTGGCAATG[A/C]TGTCTGCCTCAGATGCCCTTCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612721 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RESP18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RESP18 - regulated endocrine specific protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007089.3 | Intron | NP_001007090.3 | ||||
XM_011511165.2 | Intron | XP_011509467.1 | ||||
XM_011511167.2 | Intron | XP_011509469.1 | ||||
XM_017004079.1 | Intron | XP_016859568.1 |