Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614717 MIM: 613510 MIM: 612414 | ||||||||||||||||||||
Literature Links: |
ANAPC15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANAPC15 - anaphase promoting complex subunit 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278485.1 | 122 | Intron | NP_001265414.1 | |||
NM_001278486.1 | 122 | Intron | NP_001265415.1 | |||
NM_001278487.1 | 122 | Intron | NP_001265416.1 | |||
NM_001278488.1 | 122 | Intron | NP_001265417.1 | |||
NM_001278489.1 | 122 | Intron | NP_001265418.1 | |||
NM_001278490.1 | 122 | Intron | NP_001265419.1 | |||
NM_001278491.1 | 122 | Intron | NP_001265420.1 | |||
NM_001278492.1 | 122 | Intron | NP_001265421.1 | |||
NM_001278493.1 | 122 | Intron | NP_001265422.1 | |||
NM_001278494.1 | 122 | Intron | NP_001265423.1 | |||
NM_014042.2 | 122 | Intron | NP_054761.1 | |||
XM_017017488.1 | 122 | Intron | XP_016872977.1 | |||
XM_017017489.1 | 122 | UTR 5 | XP_016872978.1 | |||
XM_017017490.1 | 122 | Intron | XP_016872979.1 | |||
XM_017017491.1 | 122 | Intron | XP_016872980.1 | |||
XM_017017492.1 | 122 | Intron | XP_016872981.1 | |||
XM_017017493.1 | 122 | Intron | XP_016872982.1 | |||
XM_017017494.1 | 122 | Intron | XP_016872983.1 | |||
XM_017017495.1 | 122 | Intron | XP_016872984.1 | |||
XM_017017496.1 | 122 | Intron | XP_016872985.1 | |||
XM_017017497.1 | 122 | Intron | XP_016872986.1 |
LAMTOR1 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRTOMT - leucine rich transmembrane and O-methyltransferase domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |