Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611354 MIM: 615467 MIM: 601365 MIM: 605865 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPSF3L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CPSF3L - cleavage and polyadenylation specific factor 3-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256456.1 | 211 | Intron | NP_001243385.1 | |||
NM_001256460.1 | 211 | Intron | NP_001243389.1 | |||
NM_001256462.1 | 211 | Intron | NP_001243391.1 | |||
NM_001256463.1 | 211 | Intron | NP_001243392.1 | |||
NM_017871.5 | 211 | Intron | NP_060341.2 | |||
XM_011541647.1 | 211 | Intron | XP_011539949.1 | |||
XM_011541648.1 | 211 | Intron | XP_011539950.1 | |||
XM_011541650.1 | 211 | Intron | XP_011539952.1 | |||
XM_017001557.1 | 211 | Intron | XP_016857046.1 | |||
XM_017001558.1 | 211 | Intron | XP_016857047.1 |
CPTP - ceramide-1-phosphate transfer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029885.1 | 211 | Intron | NP_001025056.1 | |||
XM_005244801.3 | 211 | UTR 5 | XP_005244858.1 | |||
XM_005244802.1 | 211 | Intron | XP_005244859.1 | |||
XM_011542200.2 | 211 | Intron | XP_011540502.1 |
DVL1 - dishevelled segment polarity protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAS1R3 - taste 1 receptor member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |