Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAAGCTCTGGAGAATTTTCATAG[C/G]CAAATTCTTCTGTATGAGCTGGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606934 MIM: 612818 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDUFAF1 - NADH:ubiquinone oxidoreductase complex assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016013.3 | 1269 | Missense Mutation | GCC,GGC | A,G 314 | NP_057097.2 | |
XM_006720555.2 | 1269 | Missense Mutation | GCC,GGC | A,G 314 | XP_006720618.1 | |
XM_011521658.1 | 1269 | Missense Mutation | GCC,GGC | A,G 314 | XP_011519960.1 | |
XM_011521659.2 | 1269 | UTR 3 | XP_011519961.1 | |||
XM_017022293.1 | 1269 | Intron | XP_016877782.1 |
NUSAP1 - nucleolar and spindle associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |