Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164012 MIM: 602327 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFKB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NFKB2 - nuclear factor kappa B subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077494.3 | 3471 | Intron | NP_001070962.1 | |||
NM_001261403.2 | 3471 | Intron | NP_001248332.1 | |||
NM_001288724.1 | 3471 | Intron | NP_001275653.1 | |||
NM_001322934.1 | 3471 | Intron | NP_001309863.1 | |||
NM_001322935.1 | 3471 | Intron | NP_001309864.1 | |||
NM_002502.5 | 3471 | Intron | NP_002493.3 | |||
XM_011539830.2 | 3471 | Intron | XP_011538132.1 | |||
XM_011539831.2 | 3471 | Intron | XP_011538133.1 | |||
XM_017016278.1 | 3471 | Intron | XP_016871767.1 |
PSD - pleckstrin and Sec7 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270965.1 | 3471 | UTR 3 | NP_001257894.1 | |||
NM_001270966.1 | 3471 | UTR 3 | NP_001257895.1 | |||
NM_002779.4 | 3471 | UTR 3 | NP_002770.3 | |||
XM_011539968.1 | 3471 | UTR 3 | XP_011538270.1 | |||
XM_011539969.2 | 3471 | UTR 3 | XP_011538271.1 | |||
XM_017016433.1 | 3471 | UTR 3 | XP_016871922.1 | |||
XM_017016434.1 | 3471 | UTR 3 | XP_016871923.1 |
Set Membership: |
HapMap Validated |