Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF527 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF527 - zinc finger protein 527 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032453.1 | Intron | NP_115829.1 | ||||
XM_005259328.4 | Intron | XP_005259385.1 | ||||
XM_005259329.3 | Intron | XP_005259386.1 | ||||
XM_017027380.1 | Intron | XP_016882869.1 | ||||
XM_017027381.1 | Intron | XP_016882870.1 |