Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATAAATGGGCGGGGCCTCGGGGA[A/G]GTATGGAAACTGAAAAGAAATCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf33 - chromosome 4 open reading frame 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099783.1 | Intron | NP_001093253.1 | ||||
NM_173487.2 | Intron | NP_775758.2 | ||||
XM_005262734.4 | Intron | XP_005262791.1 | ||||
XM_017007722.1 | Intron | XP_016863211.1 | ||||
XM_017007723.1 | Intron | XP_016863212.1 | ||||
XM_017007724.1 | Intron | XP_016863213.1 |
SCLT1 - sodium channel and clathrin linker 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |