Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTTGCCACATCCTTCAGAGTGC[A/G]GCAGTAAATTCAGAAAGTATCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608991 MIM: 601733 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAML3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAML3 - mastermind like transcriptional coactivator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018717.4 | 889 | UTR 3 | NP_061187.2 |
MGST2 - microsomal glutathione S-transferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204366.1 | 889 | Intron | NP_001191295.1 | |||
NM_001204367.1 | 889 | Intron | NP_001191296.1 | |||
NM_001204368.1 | 889 | Intron | NP_001191297.1 | |||
NM_002413.4 | 889 | Intron | NP_002404.1 | |||
XM_006714221.2 | 889 | Intron | XP_006714284.1 | |||
XM_011531972.2 | 889 | Intron | XP_011530274.1 | |||
XM_017008211.1 | 889 | Intron | XP_016863700.1 | |||
XM_017008212.1 | 889 | Intron | XP_016863701.1 | |||
XM_017008213.1 | 889 | UTR 3 | XP_016863702.1 | |||
XM_017008214.1 | 889 | Intron | XP_016863703.1 |
Set Membership: |
HapMap |